ContactUsLinkComponent
Sodium hydroxide, micropearls for analysis
Corrosive
Sodium hydroxide, micropearls for analysis
Certificates
undefined
Corrosive
Sodium hydroxide, micropearls for analysis
Specification Test Results
  • Appearance:
    White Pearls
  • Titration with HCl:
    ≥99.0 %
  • Sodium carbonate (Na2CO3):
    ≤0.4 %
  • Sodium sulfate (Na2SO4):
    ≤80 ppm
  • Sodium chloride (NaCl):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Lead (Pb):
    ≤0.5 ppm