Gaskets for SVL threads, BOLA
Gasket, made of PTFE/Silicone, for tubing, tubes or probes inserted through Torion threads.
- Silicone ring with double sided PTFE
- Suitable for Torion / SVL threaded hoses
- Temperature resistance: −200 to +250 °C
Frequently Bought Together