Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Conductivity 25 °C:Max. 2.0 µS/cm
- Non-volatile matter:Max. 1 ppm
- Silver (Ag):Max. 1 ppb
- Aluminium (Al):Max. 1 ppb
- Arsenic (As):Max. 1 ppb
- Gold (Au):Max. 1 ppb
- Barium (Ba):Max. 1 ppb
- Beryllium (Be):Max. 1 ppb
- Bismuth (Bi):Max. 1 ppb
- Calcium (Ca):Max. 5 ppb
- Cadmium (Cd):Max. 1 ppb
- Cerium (Ce):Max. 1 ppb
- Cobalt (Co):Max. 1 ppb
- Chromium (Cr):Max. 1 ppb
- Cesium (Cs):Max. 1 ppb
- Copper (Cu):Max. 1 ppb
- Iron (Fe):Max. 5 ppb
- Gallium (Ga):Max. 1 ppb
- Germanium (Ge):Max. 1 ppb
- Mercury (Hg):Max. 1 ppb
- Indium (In):Max. 1 ppb
- Iridium (Ir):Max. 1 ppb
- Potassium (K):Max. 5 ppb
- Lithium (Li):Max. 1 ppb
- Magnesium (Mg):Max. 1 ppb
- Manganese (Mn):Max. 1 ppb
- Molybdenum (Mo):Max. 1 ppb
- Sodium (Na):Max. 5 ppb
- Nickel (Ni):Max. 1 ppb
- Lead (Pb):Max. 1 ppb
- Palladium (Pd):Max. 1 ppb
- Platinum (Pt):Max. 1 ppb
- Rubidium (Rb):Max. 1 ppb
- Rhodium (Rh):Max. 1 ppb
- Ruthenium (Ru):Max. 1 ppb
- Antimony (Sb):Max. 1 ppb
- Selenium (Se):Max. 1 ppb
- Tin (Sn):Max. 1 ppb
- Strontium (Sr):Max. 1 ppb
- Titanium (Ti):Max. 1 ppb
- Thallium (Tl):Max. 1 ppb
- Uran (U):Max. 0.5 ppb
- Vanadium (V):Max. 1 ppb
- Zinc (Zn):Max. 1 ppb
- Zirconium (Zr):Max. 1 ppb
- Silicon (as SiO₂):Max. 0.01 ppm
- Bromide (Br):Max. 10.0 ppb
- Chloride (Cl):Max. 10.0 ppb
- Fluoride (F):Max. 10.0 ppb
- Iodide (I):Max. 10.0 ppb
- Nitrite (NO₂):Max. 10.0 ppb
- Nitrate (NO₃):Max. 10.0 ppb
- Phosphate (PO₄):Max. 10.0 ppb
- Sulfate (SO₄):Max. 10.0 ppb
- Ammonium (NH₄):Max. 0.010 ppm
Specification Test Results
Frequently Bought Together