Water, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
Specification Test Results
- Additional info:residue (at bottling) : < 0.0005%
- Color scale:≤10 APHA
- GC/ECD (as lindane):≤5 ng/l
- Residue after evaporation:≤0.0002 %
- Acidity (CH3COOH):≤0.002 %
Specification Test Results
Frequently Bought Together