Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Potassium nitrate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 
 
Potassium nitrate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 

 

  • Assay:
    Min. 99.0 %
  • pH (5 %, 25 °C):
    5.0 - 8.0
  • Water insoluble matter:
    Max. 0.005 %
  • Ammonium (NH₄):
    Max. 0.001 %
  • Calcium (Ca):
    Max. 0.001 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 2 ppm
  • Magnesium (Mg):
    Max. 0.002 %
  • Sodium (Na):
    Max. 0.005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Iodate (IO₃):
    Max. 5 ppm
  • Nitrite (NO₂):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.002 %

 

 

Frequently Bought Together