Zadání objednávky
Czech Republic
ContactUsLinkComponent
Dusičnan cesný ≥99%, EMSURE® p.a., Supelco®
Oxidizing
Irritant
Dusičnan cesný ≥99%, EMSURE® p.a., Supelco®
Supplier:  Merck
Certificates
Oxidizing
Irritant
Dusičnan cesný ≥99%, EMSURE® p.a., Supelco®
Supplier:  Merck

Specification Test Results

  • Assay (acidimetric after ion exchange):
    ≥ 99 %
  • Chloride (Cl):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • Fe (Iron):
    ≤ 0.001 %

Specification Test Results

Related Information

Frequently Bought Together