

Sodium azide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:26628-22-8
:
Formula:NaN₃
MDL Number:MFCD00003536
:65.01 g/mol
Merck Index:13,08653
Melting Pt:275 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:247-852-1
UN:1687
ADR:6.1,II
:GHS06
:GHS09
Frequently Bought Together