


Potassium permanganate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Merck Index:12,07824
Melting Pt:> 240 °C
Density:2.7 g/cm³ (20 °C)
EINECS:231-760-3
UN:1490
ADR:5.1,II
:GHS03
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.02 %
- Pb (Lead):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
spec_VWRC26906.290 spec_VWRC26906.290 spec_VWRC26906.368 spec_VWRC26906.368
Product specification
Frequently Bought Together