Order Entry
Czech Republic
ContactUsLinkComponent
 
Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS for the determination of mercury, J.T.Baker®
  3227-01
 :  Avantor
 :  
undefined
Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS for the determination of mercury, J.T.Baker®
  3227-01
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle, amber, wide mouth, black T/E cap
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay (KMnO₄):
    ≥99.0 %
  • ACS - Insoluble Matter:
    ≤0.2 %
  • ACS - Chloride and Chlorate (as Cl):
    ≤0.005 %
  • ACS - Sulfate (SO₄):
    ≤0.02 %
  • Trace Impurities - Mercury (Hg):
    ≤0.050 ppm

 

 

 :
 :158.03 g/mol
 :12,07824
 :>240 °C
 :2.7 g/cm³ (20 °C)
 :Ambient
 :231-760-3
 :1490
 :5.1,II
 :Management of Change (MOC) category = N
 :GHS03
 :GHS07
 :GHS09
 :7722-64-7
 :KMnO₄
 :MFCD00011364