


Potassium permanganate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Merck Index:14,7655
Melting Pt:240 °C (dec.)
Density:2.7
EINECS:231-760-3
UN:1490
ADR:5.1,II
:GHS03
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together