Order Entry
Czech Republic
ContactUsLinkComponent
Periodic acid ≥99.0% (RT), crystallised, Puriss. p.a. ACS for oxidimetric titration, Fluka™
Periodic acid ≥99.0% (RT), crystallised, Puriss. p.a. ACS for oxidimetric titration, Fluka™
Catalog # 77310-100G
CAS Number:  
undefined
Periodic acid ≥99.0% (RT), crystallised, Puriss. p.a. ACS for oxidimetric titration, Fluka™
Catalog # 77310-100G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Residue on ignition (SO₄):
    Max. 0.01 %
  • Water insoluble matter:
    Max. 0.01 %
  • Silver (Ag):
    Max. 5 ppm
  • Aluminium (Al):
    Max. 5 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 30 ppm
  • Potassium (K):
    Max. 50 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 10 ppm
  • Strontium (Sr):
    Max. 5 ppm
  • Thallium (Tl):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 0.005 %
  • Iodide (I):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Other halogens (as Cl):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :227.94 g/mol
Merck Index:13,07248
Melting Pt:122 °C
Density:1,3875 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-937-0
UN:3085
ADR:5.1,II
 :GHS03
 :GHS05
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344