Periodic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10450-60-9
:
Formula:H₅IO₆
MDL Number:MFCD00011344
:227.94 g/mol
Merck Index:13,07248
Melting Pt:122 °C
Density:1.3875 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-937-0
UN:3085
ADR:5.1 (8),II
:GHS03
:GHS05
Specification Test Results
- Assay:Min. 99.5 %
- Insolubility in water:Max. 50 ppm
- Ignition residue (SO4):Max. 0.2 %
- Cl + ClO3 + Br + BrO3 (as Cl):Max. 0.02 %
- SO4 (Sulphate):Max. 100 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC20593.151 spec_VWRC20593.151 spec_VWRC20593.180
Product specification
Frequently Bought Together