Order Entry
Czech Republic
ContactUsLinkComponent
Periodic acid ≥97%, GPR RECTAPUR®
Periodic acid ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Periodic acid ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %
  • Ignition residue (SO4):
    Max. 0.5 %

Specification Test Results

Related Information

Frequently Bought Together