Order Entry
Czech Republic
ContactUsLinkComponent
Periodic acid ≥99.5% (by iodometry), EMSURE® for analysis, Supelco®
Periodic acid ≥99.5% (by iodometry), EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Periodic acid ≥99.5% (by iodometry), EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    2430 g/l (20 °C)
  • Melting Point:
    122 °C
  • Molar Mass:
    227.94 g/mol
  • Bulk Density:
    1400 kg/m3
  • pH:
    1.2 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together