Order Entry
Czech Republic
ContactUsLinkComponent
Potassium iodate ≥99%, GPR RECTAPUR®
Potassium iodate ≥99%, GPR RECTAPUR®
Catalog # 26839.264
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium iodate ≥99%, GPR RECTAPUR®
Catalog # 26839.264
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Br + Cl + ClO3 (as Cl):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :214 g/mol
Merck Index:13,07726
Melting Pt:560 °C
Density:3.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-831-9
UN:1479
ADR:5.1,II
 :GHS03
 :GHS07
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406