
Sodium iodide, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 0294.0500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:99.0 - 100.5 %
- Alkalinity:passes test
- Appearance of solution:passes test
- Iron (Fe):max. 20 ppm
- Potassium (K):passes test
- Heavy Metals (as Pb):max. 10 ppm
- Identification:passes test
- Iodate (IO3):passes test
- Limit of nitrate, nitrite and ammonia:passes test
- Loss on Drying:max. 3.0 %
- Organic Volatile Impurities:passes test
- Sulfate (SO4):max. 150 ppm
- Thiosulfate and Barium:passes test
- Thiosulfates (as S2O3):passes test
- Water (H2O):max. 2.0 %
Specification Test Results
About this item
:
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
:GHS09
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532