



Sodium cyanotrihydroborate for synthesis, Sigma-Aldrich®
8.18053.0025
: Merck
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Solubility:2100 g/l (20 °C) (slow decomposition)
- Melting Point:241 - 243 °C
- Molar Mass:62.84 g/mol
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
:
:62.84 g/mol
:13,08680
:241…243 °C
:1.11…1.12 g/cm³ (28 °C)
:Ambient
:247-317-2
:3134
:4.3 (6.1),I
:M-Clarity™ quality level = MQ200
:GHS02
:GHS06
:GHS05
:GHS09
:25895-60-7
:CH₃BNNa
:MFCD00003516