



Sodium cyanotrihydroborate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25895-60-7
:
Formula:CH₃BNNa
MDL Number:MFCD00003516
:62.84 g/mol
Merck Index:13,08680
Melting Pt:>242 °C (dec.)
Density:1,11 to 1.12 g/cm³ (28 °C)
EINECS:247-317-2
UN:3134
ADR:4.3,II
:GHS06
:GHS05
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration:≥94.0 %
- GC:≥94.0 %
Specification Test Results
Frequently Bought Together