ContactUsLinkComponent
Sodium chloride
  USBIS5000-1-1KG
 :  
Sodium chloride
  USBIS5000-1-1KG
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
 :
 :58.44 g/mol
 :14,08599
 :1413 °C (1013 hPa)
 :801 °C
 :2.16 g/cm³ (20 °C)
 :Ambient
 :231-598-3
 :7647-14-5
 :NaCl
 :MFCD00003477