Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Sodium chloride 99.0-100.5% (dried basis), powder, GenAR® USP, Macron Fine Chemicals™
 
undefined
 
Sodium chloride 99.0-100.5% (dried basis), powder, GenAR® USP, Macron Fine Chemicals™
 

 

  • CAUTION: For Manufacturing, processing or repackaging:
    99.0 - 100.5 %
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (NaCl) (dried basis):
    99.0 - 100.5 %
  • USP - Appearance of Solution:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Acidity or Alkalinity:
    Passes Test
  • USP - Iodide (I):
    Passes Test
  • USP - Bromide (Br):
    ≤100 ppm
  • USP - Loss on Drying at 105°C:
    ≤0.5 %
  • USP - Aluminum (Al)(µg/g):
    ≤0.2
  • USP - Barium (Ba):
    Passes Test
  • USP - Magnesium and Alkaline Earth Metals (as Ca):
    ≤100 ppm
  • USP - Arsenic (As):
    ≤1 ppm
  • USP - Ferrocyanide:
    Passes Test
  • USP - Iron (Fe):
    ≤2 ppm
  • USP - Potassium (K):
    ≤500 ppm
  • USP - Sulfate (SO₄):
    ≤200 ppm
  • USP - Nitrite (NO₂) (Absorbance Value):
    ≤0.01
  • USP - Phosphate (PO₄):
    ≤25 ppm
  • Solubility (5 in 50):
    Passes Test
  • 100% Passing through a US 20 Mesh Sieve:
    Passes Test
  • On U.S. No. 80 Sieve:
    30 - 50 %
  • EP/BP - Assay (NaCl) (dried basis):
    99.0 - 100.5 %
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Aluminum (Al):
    ≤0.2 ppm
  • EP/BP - Bromide (Br):
    ≤100 ppm
  • EP/BP - Barium (Ba):
    Passes Test
  • EP/BP - Iodide (I):
    Passes Test
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Loss on Drying at 100-105°C:
    ≤0.5 %
  • EP/BP - Sulfate (SO₄):
    ≤200 ppm
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Magnesium and Alkaline Earth Metals (as Ca):
    ≤100 ppm
  • EP/BP - Arsenic (As):
    ≤1 ppm
  • EP/BP - Endotoxin Concentration, <5 IU/g:
    Passes Test
  • EP/BP - Iron (Fe):
    ≤2 ppm
  • EP/BP - Ferrocyanide:
    Passes Test
  • EP/BP - Nitrite (NO₂):
    ≤0.01
  • EP/BP - Potassium (K):
    ≤500 ppm
  • EP/BP- Limit of Phosphate:
    ≤25 ppm
  • JP - Assay (NaCl) (dried basis):
    99.0 - 100.5 %
  • JP - Acidity or Alkalinity:
    Passes Test
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Barium (Ba):
    Passes Test
  • JP - Bromide (Br):
    Passes Test
  • JP - Ferrocyanide:
    Passes Test
  • JP - Iron (Fe):
    Passes Test
  • JP - Iodide (I):
    Passes Test
  • JP - Magnesium and Alkaline Earth Metals:
    Passes Test
  • JP - Phosphate (PO₄):
    Passes Test
  • JP - Sulfates (as SO₄):
    Passes Test
  • JP - Clarity and Color:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤3 ppm
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤0.5 %

 

Frequently Bought Together