Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Frequently Bought Together