Sodium cyanide, Supelco®
Catalog # 1.06437.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:13,08679
Boiling Pt:1496 °C (1013 hPa)
Melting Pt:563.9 °C
Density:1.595 g/cm³ (20 °C)
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:49.01 g/mol
EINECS:205-599-4
UN:1689
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:143-33-9
Formula:CNNa
MDL Number:MFCD00003523