Zadání objednávky
Czech Republic
ContactUsLinkComponent
Kyanid sodný 95+% ACS
Kyanid sodný 95+% ACS
Catalog # ACRO424300050
CAS Number:  
undefined
Kyanid sodný 95+% ACS
Catalog # ACRO424300050
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle for solids
  • Bal.:
    5 g
  • Sekvence:
    3173

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or flakes and/or chunks
  • Titration Argentometric:
    ≥95.0 %
  • Chloride (Cl):
    ≤1500 ppm
  • Thiocyanate (SCN):
    ≤200 ppm
  • Phosphate (PO4):
    ≤200 ppm
  • Sulfate (SO4):
    ≤500 ppm
  • Lead (Pb):
    ≤5 ppm
  • Sulfur (S):
    ≤50 ppm
  • Iron, total (as Fe):
    ≤0.005 %

Specification Test Results

About this item

 :
Merck Index:13,08679
Boiling Pt:1496 °C (1013 hPa)
Melting Pt:562 °C
Density:1,595 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-599-4
UN:1689
ADR:6.1,I
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:143-33-9
Formula:CNNa
MDL Number:MFCD00003523