Order Entry
Czech Republic
ContactUsLinkComponent
Sodium bromide, Suprapur® for trace analysis, Supelco®
Sodium bromide, Suprapur® for trace analysis, Supelco®
Katalogové číslo 1.06363.0100
Supplier:  Merck
CAS Number:  
Sodium bromide, Suprapur® for trace analysis, Supelco®
Katalogové číslo 1.06363.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    905 g/l (20 °C)
  • Melting Point:
    755 °C
  • Molar Mass:
    102.89 g/mol
  • Bulk Density:
    1750 kg/m3
  • Boiling Point:
    1393 °C
  • Vapor Pressure:
    1.3 hPa (806 °C)
  • Density:
    3.20 g/cm3 (20 °C)
  • pH:
    5.4 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :102.89 g/mol
Merck Index:13,08666
Boiling Pt:1396 °C (1013 hPa)
Melting Pt:753 °C
Density:3.2 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-599-9
CAS Number:7647-15-6
Formula:NaBr
MDL Number:MFCD00003475