ContactUsLinkComponent
Sodium bromide, anhydrous 99+%, extra pure
Sodium bromide, anhydrous 99+%, extra pure
Certificates
undefined
Sodium bromide, anhydrous 99+%, extra pure
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Titration Argentometric:
    ≥99.0 % (on dry substance)
  • Loss on drying:
    ≤1.0 %
  • Heavy metals (as Pb):
    ≤10 ppm