Potassium bromide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Katalogové číslo 1.04905.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Solubility:650 g/l (20 °C)
- Melting Point:730 °C
- Molar Mass:119.00 g/mol
- Bulk Density:900 - 1000 kg/m3
- Boiling Point:1435 °C
- Vapor Pressure:1.3 hPa (795 °C)
- Density:2.75 g/cm3 (20 °C)
- pH:5.5 - 8.5 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
:GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358