Potassium bromide, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7758-02-3
:
Formula:KBr
MDL Number:MFCD00011358
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:119 g/mol
EINECS:231-830-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:650 g/l (20 °C)
- Melting Point:730 °C
- Molar Mass:119.00 g/mol
- Bulk Density:900 - 1000 kg/m3
- Boiling Point:1435 °C
- Vapor Pressure:1.3 hPa (795 °C)
- Density:2.75 g/cm3 (20 °C)
- pH:5.5 - 8.5 (50 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.04904.0500_TDS_EN ProductPage_104904
Product specification
Merck Suprapur® High Purity Salts for Elemental Trace Analysis
Product Literature
msds_104904
SDS
Frequently Bought Together