



Sodium borohydride for synthesis, Sigma-Aldrich®
: Merck
:16940-66-2
:
:NaBH₄
:MFCD00003518
:37.83 g/mol
:13,08664
:400 °C
:1.07 g/cm³ (20 °C)
:69 °C
:1426
:4.3,I
:M-Clarity™ quality level = MQ200
:241-004-4
:GHS02
:GHS06
:GHS05
:GHS08
:Ambient
- Solubility:550 g/l (25 °C) (slow decomposition)
- Melting Point:400 °C (slow decomposition)
- Molar Mass:37.83 g/mol
- Bulk Density:350 - 500 kg/m3
- Boiling Point:500 °C (decomposition)
- Explosion Limit:3.02 %(V)
- Density:1.07 g/cm3 (20 °C)
- pH:11 (10 g/l, H2O, 20 °C)
- Ignition Point:220 °C
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
Frequently Bought Together