Order Entry
Czech Republic
ContactUsLinkComponent
Sodium borohydride 98%, granules ∼ 3 mm
Toxic
Corrosive
Flammable
Health
Sodium borohydride 98%, granules ∼ 3 mm
Certificates
undefined
Toxic
Corrosive
Flammable
Health
Sodium borohydride 98%, granules ∼ 3 mm

Specification Test Results

  • Appearance:
    White Granules
  • Titration Iodometric:
    ≥97.5 %
  • Water:
    ≤0.2 % (Coulometric)

Specification Test Results

Related Information

Frequently Bought Together