Zadání objednávky
Czech Republic
ContactUsLinkComponent
Tetrahydroboritan sodný 98+%, powder
Toxic
Corrosive
Flammable
Health
Tetrahydroboritan sodný 98+%, powder
Certificates
undefined
Toxic
Corrosive
Flammable
Health
Tetrahydroboritan sodný 98+%, powder
Certificates
Specification Test Results
  • Appearance:
    White Powder
  • Titration Iodometric:
    ≥98.0 %
  • Water:
    ≤5 %
  • HPLC:
    ≥98.5 %