Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dioxane ≥99.8% unstabilised, HiPerSolv CHROMANORM® for HPLC
Flammable
Health
Irritant
1,4-Dioxane ≥99.8% unstabilised, HiPerSolv CHROMANORM® for HPLC
Catalog # 83628.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
1,4-Dioxane ≥99.8% unstabilised, HiPerSolv CHROMANORM® for HPLC
Catalog # 83628.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.80 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 0.0005 %
  • Water:
    Max. 0.0500 %
  • Transmittance (245 nm):
    Min. 50.0 %
  • Transmittance (270 nm):
    Min. 80.0 %
  • Transmittance (300 nm):
    Min. 98.0 %
  • Conforms to BDH 15290:
    Passes test

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03330
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:204-661-8
UN:1165
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :GHS02
 :GHS08
 :GHS07
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571