
1,5-Diazobicyclo[4.3.0]-5-nonene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3001-72-7
:
Formula:C₇H₁₂N₂
MDL Number:MFCD00005554
:124.19 g/mol
Boiling Pt:110 °C (20 hPa)
Density:1 g/cm³ (20 °C)
Flash Pt:94 °C
EINECS:221-087-3
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:3267
:GHS05
Storage Temperature:Ambient
Frequently Bought Together