Order Entry
Czech Republic
ContactUsLinkComponent
 
2-Methyl-1,3-cyclopentanedione 98%
 
undefined
2-Methyl-1,3-cyclopentanedione 98%

 

  • Appearance:
    Light beige to tan Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    212 to 216 °C
  • GC:
    ≥97.5 %
  • Titration with NaOH:
    ≥97.5 %

 

 

Frequently Bought Together