

1-Ethynylcyclohexanol 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-27-3
:
Formula:C₈H₁₂O
MDL Number:MFCD00003858
:124.18 g/mol
Boiling Pt:180 °C (760 mmHg)
Melting Pt:30…32 °C
Density:0.9763 g/cm³
Flash Pt:73 °C
Storage Temperature:Ambient
EINECS:241-385-7
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4810 to 1.4840 (20°C, 589 nm)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together