
Cyclopentane
Catalog # PROOCHE142
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
Merck Index:13,02769
Boiling Pt:49 °C (1013 hPa)
Melting Pt:–93.3 °C
Density:0.746 g/cm³ (20 °C)
Flash Pt:–41 °C
Storage Temperature:Ambient
EINECS:206-016-6
UN:1146
ADR:3,II
:70.13 g/mol
:GHS02
CAS Number:287-92-3
Formula:C₅H₁₀
MDL Number:MFCD00001356