Order Entry
Czech Republic
ContactUsLinkComponent
trans-Bis(triphenylphosphine)palladium(II) chloride ≥99%
trans-Bis(triphenylphosphine)palladium(II) chloride ≥99%
Catalog # ABCRAB116703.0001
Supplier:  abcr
CAS Number:  
trans-Bis(triphenylphosphine)palladium(II) chloride ≥99%
Catalog # ABCRAB116703.0001
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :701.91 g/mol
CAS Number:13965-03-2
Formula:C₃₆H₃₀Cl₂P₂Pd
MDL Number:MFCD00009593