3-Chloropropyldimethylammonium chloride, Sigma-Aldrich®
Katalogové číslo SIALD145203-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:158.07 g/mol
:M-Clarity™ quality level = MQ200
EINECS:226-467-2
CAS Number:5407-04-5
Formula:C₅H₁₃Cl₂N
MDL Number:MFCD00012521