Methyl-2,6-difluorobenzoate, Sigma-Aldrich®
Catalog # SIAL542636-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:172.13 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:13671-00-6
Formula:C₈H₆F₂O₂
MDL Number:MFCD00051778