Order Entry
Czech Republic
ContactUsLinkComponent
Cyclohexane ≥99.7%, SPECTRONORM® for UV spectrophotometry
Cyclohexane ≥99.7%, SPECTRONORM® for UV spectrophotometry
Catalog # 23225.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Cyclohexane ≥99.7%, SPECTRONORM® for UV spectrophotometry
Catalog # 23225.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    ≥ 99.7 %
  • Density (20/4):
    0.778 - 0.779
  • Evaporation residue:
    ≤ 10 ppm
  • Water:
    ≤ 100 ppm
  • Transmittance (210 nm):
    ≥ 15 %
  • Transmittance (220 nm):
    ≥ 45 %
  • Transmittance (230 nm):
    ≥ 75 %
  • Transmittance (240 nm):
    ≥ 90 %
  • Transmittance (from 250 nm):
    ≥ 98 %

Specification Test Results

About this item

 :
 :84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119463273-41
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814