Barium trifluoromethanesulphonate, Sigma-Aldrich®
Catalog # SIAL419087-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:6.1,III
:435.45 g/mol
EINECS:220-526-6
UN:1564
CAS Number:2794-60-7
Formula:C₂BaF₆O₆S₂
MDL Number:MFCD00143644