1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
Catalog # SIAL40963-5G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:422.41 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:223437-11-4
Formula:C₁₁H₂₀F₆N₂O₄S₂
MDL Number:MFCD07784447