Brij® 52, Sigma-Aldrich®
Catalog # SIAL388831-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:330.55 g/mol
:M-Clarity™ quality level = MQ200
EINECS:500-014-1
CAS Number:9004-95-9
Formula:C₂₀H₄₂O₃