Magnesium methoxide 6 - 10% in methanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-88-6
:
Formula:C₂H₆MgO₂
MDL Number:MFCD00053671
ADR:3,II
:M-Clarity™ quality level = MQ200
:86.37 g/mol
EINECS:203-715-8
UN:1230
Frequently Bought Together