Chloro(di-tert-butyl)phosphine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13716-10-4
:
Formula:C₈H₁₈ClP
MDL Number:MFCD00008815
:180.66 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265
Frequently Bought Together