



Sodium cyanotrihydroborate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25895-60-7
:
Formula:CH₃BNNa
MDL Number:MFCD00003516
:62.84 g/mol
Merck Index:13,08680
Melting Pt:241…243 °C
Density:1.11…1.12 g/cm³ (28 °C)
Storage Temperature:Ambient
EINECS:247-317-2
ADR:4.3,II
:M-Clarity™ quality level = MQ200
UN:3134
:GHS02
:GHS06
:GHS05
:GHS09
Frequently Bought Together