1,1'-[Oxybis(ethyleneoxy)]diethylene, Sigma-Aldrich®
Catalog # SIAL139548-50ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
Density:0.97 g/cm³ (20 °C)
:158.2 g/mol
EINECS:212-133-3
CAS Number:764-99-8
Formula:H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MDL Number:MFCD00054610