Methyl-2-furoate, Sigma-Aldrich®
Catalog # SIAL129852-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:126.11 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:210-254-6
UN:2810
CAS Number:611-13-2
Formula:C₆H₆O₃
MDL Number:MFCD00003236