
1-Dodecylpyridinium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:104-74-5
:
Formula:C₁₇H₃₀ClN
MDL Number:MFCD00011987
:283.88 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:203-232-2
UN:2811
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:600 g/l (20°C)
- Melting Point:87 - 89 °C
- Flash Point:160 °C
- Density:1.04 g/cm3 (20 °C)
- pH:4 (10 g/l, H2O, 20 °C)
- Ignition Point:270°C
Specification Test Results
Frequently Bought Together