Organic reference standard, Dimethyl phthalate 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM67831.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:131-11-3
- Description:Dimethyl phthalate 100 µg/ml in n-nonane
- Formula:C₁₀H₁₀O₄
- Description:Dimethyl phthalate