Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Bis(2-ethylhexyl) adipate 100 µg/ml in n-nonane, CIL
Organic reference standard, Bis(2-ethylhexyl) adipate 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM65661.2
CAS Number:  
Organic reference standard, Bis(2-ethylhexyl) adipate 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM65661.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    103-23-1
  • Description:
    Bis(2-ethylhexyl) adipate 100 µg/ml in n-nonane
  • Formula:
    C₂₂H₄₂O₄
  • Description:
    Bis(2-ethylhexyl) adipate

Specifications

About this item